missing translation for 'onlineSavingsMsg'
Läs mer

N-(6-Aminohexyl)-2-naphthalenesulfonamide Hydrochloride, TRC

Produktkod. 30313490
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30313490 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30313490 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61105025MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material N-(6-Aminohexyl)-2-naphthalenesulfonamide, Hydrochloride
CAS 35517-14-7
Molekylformel C16 H22 N2 O2 S . Cl H
Kvantitet 25 mg
InChI formel InChI=1S/C16H22N2O2S.ClH/c17-11-5-1-2-6-12-18-21(19,20)16-10-9-14-7-3-4-8-15(14)13-16;/h3-4,7-10,13,18H,1-2,5-6,11-12,17H2;1H
LEDER Cl.NCCCCCCNS(=O)(=O)c1ccc2ccccc2c1
IUPAC-namn N-(6-aminohexyl)naphthalene-2-sulfonamide;hydrochloride
Molekylvikt (g/mol) 342.88
Formel vikt 342.1169
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.