Läs mer
N-(6-Aminohexyl)-N'-(4-chlorophenyl)imidodicarbonimidic Diamide Dihydrochloride, TRC
Produktkod. 30135783
Change view
Kvantitet:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135783 50 mg
30133177 250 mg
17993085 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135783 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61005050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 165678-61-5
Molekylformel C14H25Cl3N6
Molekylvikt (g/mol) 383.75
Synonym Imidodicarbonimidic diamide, N-(6-aminohexyl)-N'-(4-chlorophenyl)-, hydrochloride (1:2), Imidodicarbonimidic diamide, N-(6-aminohexyl)-N'-(4-chlorophenyl)-, dihydrochloride (9CI), 1-(6-Aminohexyl)-5-(4-chlorophenyl)biguanide Dihydrochloride
IUPAC-namn 1-[amino-(4-chloroanilino)methylidene]-2-(6-aminohexyl)guanidine
LEDER NCCCCCCNC(NC(NC1=CC=C(Cl)C=C1)=N)=N
Specifikationer
Kemiskt namn eller material N-(6-Aminohexyl)-N'-(4-chlorophenyl)imidodicarbonimidic Diamide Dihydrochloride
Analyt- eller komponentnamn N-(6-Aminohexyl)-N'-(4-chlorophenyl)imidodicarbonimidic Diamide Dihydrochloride
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C14H23ClN6/c15-11-5-7-12(8-6-11)20-14(18)21-13(17)19-10-4-2-1-3-9-16/h5-8H,1-4,9-10,16H2,(H5,17,18,19,20,21)
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 50 mg
Formel vikt 382.1206
Procent renhet >95%