Läs mer
N-[(6-Chloropyridin-3-yl)methyl]methylamine, TRC
Produktkod. 30010014
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010014 500 mg
30005456 5 g
30006774 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010014 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC380145500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 120739-62-0
Molekylformel C7 H9 Cl N2
Molekylvikt (g/mol) 156.61
Synonym 6-Chloro-N-methyl-3-pyridinemethanamine, 1-(6-Chloropyridin-3-yl)-N-methylmethanamine, 2-Chloro-5-(methylaminomethyl)pyridine, N-(6-Chloro-3-pyridylmethyl)-N-methylamine, N-[(6-Chloropyridin-3-yl)methyl]methylamine
IUPAC-namn 1-(6-chloropyridin-3-yl)-N-methylmethanamine
LEDER CNCc1ccc(Cl)nc1
Specifikationer
Kemiskt namn eller material Acetamiprid metabolite IM-1-4
Kvantitet 500 mg
InChI formel InChI=1S/C7H9ClN2/c1-9-4-6-2-3-7(8)10-5-6/h2-3,5,9H,4H2,1H3
Formel vikt 156.0454