missing translation for 'onlineSavingsMsg'
Läs mer

N-[9-[[2-Hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6-oxo-1H-purin-2-yl]acetamide (Ganciclovir N-Acetate), Mikromol™

Product Code. 30050411
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30050411 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050411 Supplier Mikromol Supplier No. MM0485.170025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-[9-[[2-Hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6-oxo-1H-purin-2-yl]acetamide (Ganciclovir N-Acetate)
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-[9-[(2-Hydroxy-1-(hydroxymethyl)ethoxy)methyl]-6-oxo-1H-purin-2-yl]acetamide
CAS 84245-12-5
Analyt- eller komponentnamn N-[9-[(2-Hydroxy-1-(hydroxymethyl)ethoxy)methyl]-6-oxo-1H-purin-2-yl]acetamide
Frakt skick Room Temperature
Molekylformel C11 H15 N5 O5
Certifikat och efterlevnad ISO 9001
Synonym Acetamide, N-[6,9-dihydro-9-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6-oxo-1H-purin-2-yl]-, N-[6,9-Dihydro-9-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6-oxo-1H-purin-2-yl]acetamide, N-[9-[[2-Hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6-oxo-1H-purin-2-yl]acetamide, Ganciclovir N-Acetate, N-Acetylganciclovir
InChI formel InChI=1S/C11H15N5O5/c1-6(19)13-11-14-9-8(10(20)15-11)12-4-16(9)5-21-7(2-17)3-18/h4,7,17-18H,2-3,5H2,1H3,(H2,13,14,15,19,20)
LEDER CC(=O)NC1=Nc2c(ncn2COC(CO)CO)C(=O)N1
IUPAC-namn N-[9-(1,3-dihydroxypropan-2-yloxymethyl)-6-oxo-1H-purin-2-yl]acetamide
Molekylvikt (g/mol) 297.2673
Kvantitet 25 mg
Formel vikt 297.1073 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.