Läs mer
N-[9-[(2-Hydroxyethoxy)-methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide, Mikromol™
Produktkod. 17920896
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17920896 25 mg
17923476 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17920896 Leverantör Mikromol Leverantörsnummer MM0061.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[9-[(2-Hydroxyethoxy)-methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[9-[(2-Hydroxyethoxy)-methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide
CAS 110104-37-5
Analyt- eller komponentnamn N-[9-[(2-Hydroxyethoxy)-methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide
Frakt skick Room Temperature
Molekylformel C10 H13 N5 O4
Certifikat och efterlevnad ISO 17025
Synonym Aciclovir Imp. F (EP), Acyclovir USP Related Compound F, Acyclovir USP RC F, N-[9-[(2-Hydroxyethoxy)-methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide, Acyclovir Related Compound F, Aciclovir Impurity F
InChI formel InChI=1S/C10H13N5O4/c1-6(17)12-10-13-8-7(9(18)14-10)11-4-15(8)5-19-3-2-16/h4,16H,2-3,5H2,1H3,(H2,12,13,14,17,18)
LEDER CC(=O)NC1=Nc2c(ncn2COCCO)C(=O)N1
IUPAC-namn N-[9-(2-hydroxyethoxymethyl)-6-oxo-1H-purin-2-yl]acetamide
Molekylvikt (g/mol) 267.24
Kvantitet 25 mg
Formel vikt 267.0968 g/mol