Läs mer
N-Acetyl-2-O-benzyl-neuraminic Acid Methyl Ester, 85%, TRC
Produktkod. 30322872
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30322872 250 mg
1 options
Produktkod. 30322872 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA170805250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-2-O-benzyl-neuraminic Acid Methyl Ester
CAS 57777-97-6
Molekylformel C19H27NO9
Kvantitet 250 mg
Synonym Benzyl 5-(Acetylamino)-3,5-dideoxy-D-glycero-D-galacto-2-nonulopyranosidonic Acid Methyl Ester
InChI formel InChI=1S/C19H27NO9/c1-11(22)20-15-13(23)8-19(18(26)27-2,28-10-12-6-4-3-5-7-12)29-17(15)16(25)14(24)9-21/h3-7,13-17,21,23-25H,8-10H2,1-2H3,(H,20,22)/t13-,14-,15+,16+,17+,19+/m0/s1
LEDER COC(=O)[C@]1(C[C@H](O)[C@@H](NC(=O)C)[C@@H](O1)[C@H](O)[C@@H](O)CO)OCc2ccccc2
IUPAC-namn methyl (2R,4S,5R,6R)-5-acetamido-4-hydroxy-2-phenylmethoxy-6-[(1R,2S)-1,2,3-trihydroxypropyl]oxane-2-carboxylate
Molekylvikt (g/mol) 413.42
Formel vikt 413.1686