Läs mer
N-Acetyl-2-O-methyl-Alpha-neuraminic Acid Methyl Ester 4,7,8,9-Tetraacetate, TRC
Produktkod. 30320758
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30320758 50 mg
1 options
Produktkod. 30320758 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18672050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-2-O-methyl-a-neuraminic Acid Methyl Ester 4,7,8,9-T
CAS 73208-80-7
Molekylformel C21 H31 N O13
Kvantitet 50 mg
Synonym N-Acetyl-2-O-Methyl-α-Neuraminic Acid Methyl Ester 4,7,8,9-Tetraacetate
InChI formel InChI=1S/C21H31NO13/c1-10(23)22-17-15(32-12(3)25)8-21(30-7,20(28)29-6)35-19(17)18(34-14(5)27)16(33-13(4)26)9-31-11(2)24/h15-19H,8-9H2,1-7H3,(H,22,23)/t15-,16+,17+,18+,19+,21+/m0/s1
LEDER COC(=O)[C@]1(C[C@H](OC(=O)C)[C@@H](NC(=O)C)[C@@H](O1)[C@H](OC(=O)C)[C@@H](COC(=O)C)OC(=O)C)OC
IUPAC-namn methyl (2R,4S,5R,6R)-5-acetamido-4-acetyloxy-2-methoxy-6-[(1S,2R)-1,2,3-triacetyloxypropyl]oxane-2-carboxylate
Molekylvikt (g/mol) 505.47
Formel vikt 505.1795