Läs mer
N-Acetyl Axitinib, TRC
Produktkod. 30135198
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135198 10 mg
30137762 2.5 mg
30011314 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135198 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA13456010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1639137-80-6
Molekylformel C24H20N4O2S
Molekylvikt (g/mol) 428.51
Synonym (E)-2-((1-Acetyl-3-(2-(pyridin-2-yl)vinyl)-1H-indazol-6-yl)thio)-N-methylbenzamide
IUPAC-namn 2-[1-acetyl-3-[(E)-2-pyridin-2-ylethenyl]indazol-6-yl]sulfanyl-N-methylbenzamide
LEDER CNC(=O)c1ccccc1Sc2ccc3c(\C=C\c4ccccn4)nn(C(=O)C)c3c2
Specifikationer
Kemiskt namn eller material N-Acetyl Axitinib
Analyt- eller komponentnamn N-Acetyl Axitinib
Frakt skick Room Temperature
InChI formel InChI=1S/C24H20N4O2S/c1-16(29)28-22-15-18(31-23-9-4-3-8-20(23)24(30)25-2)11-12-19(22)21(27-28)13-10-17-7-5-6-14-26-17/h3-15H,1-2H3,(H,25,30)/b13-10+
Lösningstyp Neat
Industry Type Pharmaceutical