Läs mer
N-Acetyl-b-D-Glucosamine 6-Acetate 1,3,4-Tribenzyl Ether, TRC
Produktkod. 30324618
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30324618 50 mg
1 options
Produktkod. 30324618 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA17819050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-Beta-D-Glucosamine 6-Acetate 1,3,4-Tribenzyl Ether
CAS 129729-33-5
Molekylformel C31H35NO7
Kvantitet 50 mg
Synonym Phenylmethyl 2-(Acetylamino)-2-deoxy-3,4-bis-O-(phenylmethyl)-β-D-glucopyranoside 6-Acetate
InChI formel InChI=1S/C31H35NO7/c1-22(33)32-28-30(37-19-25-14-8-4-9-15-25)29(36-18-24-12-6-3-7-13-24)27(21-35-23(2)34)39-31(28)38-20-26-16-10-5-11-17-26/h3-17,27-31H,18-21H2,1-2H3,(H,32,33)/t27?,28?,29-,30?,31-/m1/s1
LEDER CC(=O)NC1[C@H](OCc2ccccc2)OC(COC(=O)C)[C@@H](OCc3ccccc3)C1OCc4ccccc4
IUPAC-namn [(3S,6R)-5-acetamido-3,4,6-tris(phenylmethoxy)oxan-2-yl]methyl acetate
Molekylvikt (g/mol) 533.61
Formel vikt 533.2414