missing translation for 'onlineSavingsMsg'
Läs mer

N-Acetyl Bisoprolol, TRC

Produktkod. 30325879
Change view
Click to view available options
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30325879 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325879 Supplier Toronto Research Chemicals Supplier No. TRCA19258810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-Acetylbisoprolol
CAS 66722-65-4
Molekylformel C20 H33 N O5
Kvantitet 10 mg
Synonym N-[2-hydroxy-3-[4-[[2-(1-methylethoxy)ethoxy]methyl]phenoxy]propyl]-N-(1-methylethyl)acetamide, N-Acetylbisoprolol
InChI formel InChI=1S/C20H33NO5/c1-15(2)21(17(5)22)12-19(23)14-26-20-8-6-18(7-9-20)13-24-10-11-25-16(3)4/h6-9,15-16,19,23H,10-14H2,1-5H3
LEDER CC(C)OCCOCc1ccc(OCC(O)CN(C(C)C)C(=O)C)cc1
IUPAC-namn N-[2-hydroxy-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propyl]-N-propan-2-ylacetamide
Molekylvikt (g/mol) 367.48
Formel vikt 367.2359
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.