Läs mer
N-Acetyl-D-alanyl-D-alanine, TRC
Produktkod. 30323218
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30323218 250 mg
1 options
Produktkod. 30323218 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA168068250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-D-alanyl-D-alanine
CAS 19993-26-1
Molekylformel C8H14N2O4
Kvantitet 250 mg
Synonym N-Acetyl-D-Ala-D-Ala-oh, Acetyl-D-alanyl-D-alanine, N-Acetyl-D-alanyl-D-alanine
InChI formel InChI=1S/C8H14N2O4/c1-4(9-6(3)11)7(12)10-5(2)8(13)14/h4-5H,1-3H3,(H,9,11)(H,10,12)(H,13,14)/t4-,5-/m1/s1
LEDER C[C@@H](NC(=O)[C@@H](C)NC(=O)C)C(=O)O
IUPAC-namn (2R)-2-[[(2R)-2-acetamidopropanoyl]amino]propanoic acid
Molekylvikt (g/mol) 202.21
Formel vikt 202.0954