Läs mer
N-Acetyl-D-allo-isoleucine, TRC
Produktkod. 30178988
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30178988 100 mg
30178979 50 mg
30179286 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30178988 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA168193100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54831-20-8
Molekylformel C8H15NO3
Molekylvikt (g/mol) 173.21
IUPAC-namn (2R,3S)-2-acetamido-3-methylpentanoic acid
LEDER CC[C@H](C)[C@@H](NC(=O)C)C(=O)O
Specifikationer
Kemiskt namn eller material N-Acetyl-D-allo-isoleucine
InChI formel InChI=1S/C8H15NO3/c1-4-5(2)7(8(11)12)9-6(3)10/h5,7H,4H2,1-3H3,(H,9,10)(H,11,12)/t5-,7+/m0/s1
Kvantitet 100 mg
Formel vikt 173.1052