Läs mer
N-Acetyl-D-galactosamine 4-Sulfate Sodium Salt, TRC
Produktkod. 30317215
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30317215 25 mg
1 options
Produktkod. 30317215 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA13276025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-D-galactosamine 4-Sulfate Sodium Salt
CAS 660839-03-2
Molekylformel C8 H14 N Na O9 S
Kvantitet 25 mg
Synonym N-Acetylgalactosamine 4-Sulfate Sodium Salt
InChI formel InChI=1S/C8H15NO9S.Na/c1-3(11)9-5-6(12)7(18-19(14,15)16)4(2-10)17-8(5)13;/h4-8,10,12-13H,2H2,1H3,(H,9,11)(H,14,15,16);/q;+1/p-1/t4-,5-,6-,7+,8?;/m1./s1
LEDER CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](OS(=O)(=O)O[Na])[C@@H]1O
IUPAC-namn [(2R,3R,4R,5R)-5-acetamido-4,6-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxysulfonyloxysodium
Molekylvikt (g/mol) 323.253
Formel vikt 323.029