Läs mer
N-Acetyl-D-Glucosamine 6-Acetate, TRC
Produktkod. 30311005
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30311005 25 mg
1 options
Produktkod. 30311005 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA17820525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-D-Glucosamine 6-Acetate
CAS 131832-93-4
Molekylformel C10H17NO7
Kvantitet 25 mg
Synonym 2-(Acetylamino)-2-deoxy-D-glucose 6-Acetate, (N,6-O)-Diacetyl-D-Glucosamine
InChI formel InChI=1S/C10H17NO7/c1-4(12)11-7-9(15)8(14)6(18-10(7)16)3-17-5(2)13/h6-10,14-16H,3H2,1-2H3,(H,11,12)/t6-,7-,8-,9-,10?/m1/s1
LEDER O[C@H]1C(O)C(NC(C)=O)[C@H](O)OC1COC(C)=O
IUPAC-namn [(2R,3S,4R,5R)-5-acetamido-3,4,6-trihydroxyoxan-2-yl]methyl acetate
Molekylvikt (g/mol) 263.24
Formel vikt 263.1005