Läs mer
N-Acetyl D,L-Alpha-Methyl DOPA Dimethyl Ether, TRC
Produktkod. 30308752
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30308752 25 mg
1 options
Produktkod. 30308752 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18665025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl D,L-alpha-Methyl DOPA Dimethyl Ether
CAS 5934-66-7
Molekylformel C14H19NO5
Kvantitet 25 mg
Synonym N-Acetyl-3-methoxy-O,alpha-dimethyl-tyrosine, N-Acetyl-3-methoxy-O,alpha-dimethyl-DL-tyrosine
InChI formel InChI=1S/C14H19NO5/c1-9(16)15-14(2,13(17)18)8-10-5-6-11(19-3)12(7-10)20-4/h5-7H,8H2,1-4H3,(H,15,16)(H,17,18)
LEDER COc1ccc(CC(C)(NC(=O)C)C(=O)O)cc1OC
IUPAC-namn 2-acetamido-3-(3,4-dimethoxyphenyl)-2-methylpropanoic acid
Molekylvikt (g/mol) 281.3
Formel vikt 281.1263