Läs mer
N-Acetyl-D,L-homotryptophan, TRC
Produktkod. 30328205
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30328205 100 mg
1 options
Produktkod. 30328205 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA178400100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-D,L-homotryptophan
CAS 205813-00-9
Molekylformel C14 H16 N2 O3
Kvantitet 100 mg
Synonym 1H-Indole-3-butanoic acid, α-(acetylamino)-, N-Acetyl-D,L-homotryptophan, N-Acetyl-DL-homotryptophan, α-(Acetylamino)-1H-indole-3-butanoic acid, 2-Acetamido-4-(1H-indol-3-yl)butanoic acid
InChI formel InChI=1S/C14H16N2O3/c1-9(17)16-13(14(18)19)7-6-10-8-15-12-5-3-2-4-11(10)12/h2-5,8,13,15H,6-7H2,1H3,(H,16,17)(H,18,19)
LEDER CC(=O)NC(CCc1c[nH]c2ccccc12)C(=O)O
IUPAC-namn 2-acetamido-4-(1H-indol-3-yl)butanoic acid
Molekylvikt (g/mol) 260.29
Formel vikt 260.1161