Läs mer
N-Acetyl-D-mannosamine, TRC
Produktkod. 30313582
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30313582 1 g
1 options
Produktkod. 30313582 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1861991G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-D-mannosamine
CAS 3615-17-6
Anteckningar om renhetsgrad HPLC
Molekylformel C8 H15 N O6
Kvantitet 1 g
Synonym D-Mannose, 2-(acetylamino)-2-deoxy-, Mannose, 2-acetamido-2-deoxy-, D- (8CI), 2-(Acetylamino)-2-deoxy-D-mannose, 2-Acetamido-2-deoxy-D-mannopyranose, 2-Acetamido-2-deoxy-D-mannose, D-N-Acetylmannosamine, D-N-Acetylmannosamine, N-Acetyl-2-amino-2-deoxy-D-mannose, N-Acetyl-D-mannosamine, N-Acetyl-β-D-mannosamine, N-Acetylmannosamine
InChI formel InChI=1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4-,5+,6-,7-,8?/m1/s1
LEDER CC(=O)N[C@@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
IUPAC-namn N-[(3S,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Molekylvikt (g/mol) 221.21
Formel vikt 221.0899