missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-phenylalanine, 95%, Thermo Scientific™

Catalog Number p-7054934
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
10172-89-1
C11H13NO3
207.23
MFCD00002664
CBQJSKKFNMDLON-SNVBAGLBSA-N
n-acetyl-d-phenylalanine, acetyl-d-phenylalanine, ac-d-phe-oh, n-acetyl-3-phenyl-d-alanine, n-acetylphenylalanine, d, d-phenylalanine, n-acetyl, r-2-acetamido-3-phenylpropanoic acid, 2r-2-acetamido-3-phenylpropanoic acid, n-acetylphenylalanine #, n-ac-d-phe-oh
101184
CHEBI:28203
(2R)-2-acetamido-3-phenylpropanoic acid
CC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O
This item is not returnable. View return policy
10172-89-1
C11H13NO3
207.23
MFCD00002664
CBQJSKKFNMDLON-SNVBAGLBSA-N
n-acetyl-d-phenylalanine, acetyl-d-phenylalanine, ac-d-phe-oh, n-acetyl-3-phenyl-d-alanine, n-acetylphenylalanine, d, d-phenylalanine, n-acetyl, r-2-acetamido-3-phenylpropanoic acid, 2r-2-acetamido-3-phenylpropanoic acid, n-acetylphenylalanine #, n-ac-d-phe-oh
101184
CHEBI:28203
(2R)-2-acetamido-3-phenylpropanoic acid
CC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 165°C to 169°C
Color White
Beilstein 2213852
Formula Weight 207.23
Percent Purity 95%
Quantity 5 g
Physical Form Solid
Chemical Name or Material N-Acetyl-D-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'