missing translation for 'onlineSavingsMsg'
Läs mer

N-Acetyl-d3-5-hydroxytryptamine, TRC

Produktkod. 30325660
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30325660 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30325660 Supplier Toronto Research Chemicals Supplier No. TRCA1791620.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-Acetyl-d3-5-hydroxytryptamine
CAS 2001098-07-1
Molekylformel C12 D3 H11 N2 O2
Kvantitet 0.5 mg
Synonym N-[2-(5-Hydroxy-1H-indol-3-yl)ethyl]acetamide-d3, 5-Hydroxy-N-acetyltryptamine-d3, 5-Hydroxymelatonin-d3, N-Acetylserotonin-d3, N-[2-(5-Hydroxy-1H-indol-3-yl)ethyl]trideuterioacetamide, 5-Hydroxymelatonin-d3, N-(Acetyl-d3)-2-(5-hydroxyindol-3-yl)ethylamine, N-(Acetyl-d3)-5-hydroxytryptamine, N-Acetyl-5-hydroxytryptamine-D3 (N-acetyl-D3), N-Acetyl-d3-serotonin, Normelatonin-d3, O-Demethylmelatonin-d3
InChI formel InChI=1S/C12H14N2O2/c1-8(15)13-5-4-9-7-14-12-3-2-10(16)6-11(9)12/h2-3,6-7,14,16H,4-5H2,1H3,(H,13,15)/i1D3
LEDER [2H]C([2H])([2H])C(=O)NCCc1c[nH]c2ccc(O)cc12
IUPAC-namn 2,2,2-trideuterio-N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]acetamide
Molekylvikt (g/mol) 221.27
Formel vikt 221.1244
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.