Läs mer
N-Acetyl-dehydro-Eletriptan, TRC
Produktkod. 30310422
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30310422 25 mg
1 options
Produktkod. 30310422 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA17228525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-dehydro-Eletriptan
CAS 188113-71-5
Molekylformel C24H26N2O3S
Kvantitet 25 mg
Synonym 1-Acetyl-3-[[(2R)-1-methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethenyl]- 1H-indole, (R,E)-1-(3-((1-methylpyrrolidin-2-yl)methyl)-5-(2-(phenylsulfonyl)vinyl) -1H-indol-1-yl)ethanone
InChI formel InChI=1S/C24H26N2O3S/c1-18(27)26-17-20(16-21-7-6-13-25(21)2)23-15-19(10-11-24(23)26)12-14-30(28,29)22-8-4-3-5-9-22/h3-5,8-12,14-15,17,21H,6-7,13,16H2,1-2H3/b14-12+/t21-/m1/s1
LEDER O=S(/C=C/C1=CC=C(N(C(C)=O)C=C2C[C@@H]3CCCN3C)C2=C1)(C4=CC=CC=C4)=O
IUPAC-namn 1-[5-[(E)-2-(benzenesulfonyl)ethenyl]-3-[[(2R)-1-methylpyrrolidin-2-yl]methyl]indol-1-yl]ethanone
Molekylvikt (g/mol) 422.54