Läs mer
N-Acetyl Demethyl Memantine, TRC
Produktkod. 17902875
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17902875 25 mg
17960535 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17902875 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA17351025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 778-09-6
Molekylformel C13 H21 N O
Molekylvikt (g/mol) 207.3119
Synonym N-(3-Methyltricyclo[3.3.1.13,7]dec-1-yl)acetamide, N-(3-Methyl-1-adamantyl)acetamide, 1-Acetamido-3-methyladamantane, 1-Methyl-3-acetamidoadamantane, NSC 179381
IUPAC-namn N-[(5R,7S)-3-methyl-1-adamantyl]acetamide
LEDER CC(=O)N[C@@]12C[C@H]3C[C@H](C[C@@](C)(C3)C1)C2
Specifikationer
Kemiskt namn eller material N-Acetyl Demethyl Memantine
Kvantitet 25 mg
InChI formel InChI=1S/C13H21NO/c1-9(15)14-13-6-10-3-11(7-13)5-12(2,4-10)8-13/h10-11H,3-8H2,1-2H3,(H,14,15)/t10-,11+,12-,13-
Formel vikt 207.1623