Läs mer
N-Acetyl-L-arginine Methyl Ester Hydrochloride, TRC
Produktkod. 30326909
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30326909 1 g
1 options
Produktkod. 30326909 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1683601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-L-arginine Methyl Ester Hydrochloride
CAS 1784-05-0
Molekylformel C9 H18 N4 O3 . Cl H
Kvantitet 1 g
Synonym N-Acetyl-L-arginine Methyl Ester Hydrochloride, N2-Acetyl-L-arginine Methyl Ester Monohydrochloride, Ac-L-Arg-OMe Hydrochloride
InChI formel InChI=1S/C9H18N4O3.ClH/c1-6(14)13-7(8(15)16-2)4-3-5-12-9(10)11;/h7H,3-5H2,1-2H3,(H,13,14)(H4,10,11,12);1H/t7-;/m0./s1
LEDER Cl.COC(=O)[C@H](CCCNC(=N)N)NC(=O)C
IUPAC-namn methyl (2S)-2-acetamido-5-carbamimidamidopentanoate;hydrochloride
Molekylvikt (g/mol) 266.725
Formel vikt 266.115