Läs mer
N-Acetyl-L-cysteine Isopropyl Ester, TRC
Produktkod. 30320494
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30320494 100 mg
1 options
Produktkod. 30320494 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA457890100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-L-cysteine Isopropyl Ester
CAS 73255-51-3
Molekylformel C8 H15 N O3 S
Kvantitet 100 mg
Synonym N-Acetyl-L-cysteine 1-methylethyl ester, Cysteine, N-acetyl-, isopropyl ester (6CI), N-Acetyl-L-cysteine isopropyl ester
InChI formel InChI=1S/C8H15NO3S/c1-5(2)12-8(11)7(4-13)9-6(3)10/h5,7,13H,4H2,1-3H3,(H,9,10)/t7-/m0/s1
LEDER CC(C)OC(=O)[C@H](CS)NC(=O)C
IUPAC-namn propan-2-yl (2R)-2-acetamido-3-sulfanylpropanoate
Molekylvikt (g/mol) 205.27
Formel vikt 205.0773