Läs mer
N-Acetyl-L-tyrosine Amide, TRC
Produktkod. 30315956
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30315956 1 g
1 options
Produktkod. 30315956 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1897401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-L-tyrosine Amide
CAS 1948-71-6
Molekylformel C11H14N2O3
Kvantitet 1 g
Synonym (alphaS)-alpha-(Acetylamino)-4-hydroxybenzenepropanamide, L-alpha-Acetamido-p-hydroxyhydrocinnamamide, N-Acetyl-4-hydroxy-L-phenylalaninamide, N-Acetyl-L-tyrosinamide, NAYA, Ac-L-Tyr-NH2
InChI formel InChI=1S/C11H14N2O3/c1-7(14)13-10(11(12)16)6-8-2-4-9(15)5-3-8/h2-5,10,15H,6H2,1H3,(H2,12,16)(H,13,14)/t10-/m0/s1
LEDER CC(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)N
IUPAC-namn (2S)-2-acetamido-3-(4-hydroxyphenyl)propanamide
Molekylvikt (g/mol) 222.24
Formel vikt 222.1004