Läs mer
N-Acetyl Memantine, TRC
Produktkod. 30138147
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138147 250 mg
17951005 500 mg
17941575 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138147 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA186250250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19982-07-1
Molekylformel C14 H23 N O
Molekylvikt (g/mol) 221.34
Synonym Acetamide, N-(3,5-dimethyltricyclo[3.3.1.13,7]dec-1-yl)-, Acetamide, N-(3,5-dimethyl-1-adamantyl)- (7CI,8CI), N-(3,5-Dimethyltricyclo[3.3.1.13,7]dec-1-yl)acetamide, 1-Acetamido-3,5-dimethyladamantane, 1-Acetylamino-3,5-dimethyladamantane, N-Acetylmemantine
IUPAC-namn N-(3,5-dimethyl-1-adamantyl)acetamide
LEDER CC(=O)NC12CC3CC(C)(CC(C)(C3)C1)C2
Specifikationer
Kemiskt namn eller material N-Acetyl Memantine
Kvantitet 250 mg
InChI formel InChI=1S/C14H23NO/c1-10(16)15-14-6-11-4-12(2,8-14)7-13(3,5-11)9-14/h11H,4-9H2,1-3H3,(H,15,16)
Formel vikt 221.178