Läs mer
N-Acetyl N-Descarboxyethyl Retigabine, TRC
Produktkod. 30328250
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30328250 25 mg
1 options
Produktkod. 30328250 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18793025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetylretigabine
CAS 229970-68-7
Molekylformel C15 H16 F N3 O
Kvantitet 25 mg
Synonym N-Acetyl Retigabine, N-Acetylretigabine
InChI formel InChI=1S/C15H16FN3O/c1-10(20)19-15-7-6-13(8-14(15)17)18-9-11-2-4-12(16)5-3-11/h2-8,18H,9,17H2,1H3,(H,19,20)
LEDER CC(=O)Nc1ccc(NCc2ccc(F)cc2)cc1N
IUPAC-namn N-[2-amino-4-[(4-fluorophenyl)methylamino]phenyl]acetamide
Molekylvikt (g/mol) 273.31
Formel vikt 273.1277