Läs mer
N-Acetyl-N-tert-butoxycarbonyl Serotonin, TRC
Produktkod. 30315500
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30315500 100 mg
1 options
Produktkod. 30315500 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA171155100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-N-tert-butoxycarbonyl Serotonin
CAS 1329837-64-0
Molekylformel C17H22N2O4
Kvantitet 100 mg
Synonym N-tert-Butoxycarbonyl-normelatonin, N-tert-Butoxycarbonyl-O-demethylmelatonin, N-[2-(N-tert-Butoxycarbonyl-5-hydroxyindol-3-yl)ethyl]-acetamide, N-tert-Butoxycarbonyl-5-hydroxymelatonin, [1-(N-tert-Butoxycarbonyl)-3-(2-acetamidoethyl)-5-hydroxy]indole
InChI formel InChI=1S/C17H22N2O4/c1-11(20)18-8-7-12-10-19(16(22)23-17(2,3)4)15-6-5-13(21)9-14(12)15/h5-6,9-10,21H,7-8H2,1-4H3,(H,18,20)
LEDER CC(=O)NCCc1cn(C(=O)OC(C)(C)C)c2ccc(O)cc12
IUPAC-namn tert-butyl 3-(2-acetamidoethyl)-5-hydroxyindole-1-carboxylate
Molekylvikt (g/mol) 318.37
Formel vikt 318.158