Läs mer
N-Acetyl-O-methyl-D-serine, TRC
Produktkod. 30320981
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30320981 50 mg
1 options
Produktkod. 30320981 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18636050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-O-methyl-D-serine
CAS 196601-67-9
Molekylformel C6H11NO4
Kvantitet 50 mg
Synonym N-Acetyl-O-methyl-D-serine, (R)-2-(Acetylamino)-3-methoxypropionic Acid
InChI formel InChI=1S/C6H11NO4/c1-4(8)7-5(3-11-2)6(9)10/h5H,3H2,1-2H3,(H,7,8)(H,9,10)/t5-/m1/s1
LEDER COC[C@@H](NC(=O)C)C(=O)O
IUPAC-namn (2R)-2-acetamido-3-methoxypropanoic acid
Molekylvikt (g/mol) 161.16
Formel vikt 161.0688