Läs mer
N-Acetyl Phentermine, TRC
Produktkod. 30483251
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483251 250 mg
30471543 100 mg
30472487 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30483251 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM332870250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5531-33-9
Molekylformel C12H17NO
Molekylvikt (g/mol) 191.27
Synonym N-(1,1-Dimethyl-2-phenylethyl)acetamide, N-(alpha,alpha-Dimethylphenethyl)acetamide, NSC 103909, N-(2-Methyl-1-phenylpropan-2-yl)acetamide
IUPAC-namn N-(2-methyl-1-phenylpropan-2-yl)acetamide
LEDER CC(=O)NC(C)(C)Cc1ccccc1
Specifikationer
Kemiskt namn eller material N-Acetyl Phentermine
Kvantitet 250 mg
InChI formel InChI=1S/C12H17NO/c1-10(14)13-12(2,3)9-11-7-5-4-6-8-11/h4-8H,9H2,1-3H3,(H,13,14)
Formel vikt 191.131