Läs mer
N-Acetyl-S-(2-ethoxycarbonylethyl-1-methyl)-L-cysteine, Dicyclohexylammonium Salt(Mixture of Diastereomers), TRC
Produktkod. 30322878
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30322878 25 mg
1 options
Produktkod. 30322878 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA17379025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-S-(2-ethoxycarbonylethyl-1-methyl)-L-cysteine, Dicyclohexylammonium Salt
CAS 33297-48-2
Molekylformel C12 H23 N . C11 H19 N O5 S
Kvantitet 25 mg
Synonym N-Acetyl-S-(2-Ethoxycarbonylethyl-1-Methyl)-L-Cysteine, Dicyclohexylammonium Salt
InChI formel InChI=1S/C12H23N.C11H19NO5S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-4-17-10(14)5-7(2)18-6-9(11(15)16)12-8(3)13/h11-13H,1-10H2;7,9H,4-6H2,1-3H3,(H,12,13)(H,15,16)/t;7?,9-/m.0/s1
LEDER CCOC(=O)CC(C)SC[C@H](NC(=O)C)C(=O)O.C1CCC(CC1)NC2CCCCC2
IUPAC-namn (2R)-2-acetamido-3-(4-ethoxy-4-oxobutan-2-yl)sulfanylpropanoic acid;N-cyclohexylcyclohexanamine
Molekylvikt (g/mol) 458.66
Formel vikt 458.2814