Läs mer
N-Acetyl-S-(2-hydroxyethyl-d4)-L-cysteine Dicyclohexylamine Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Acetyl-S-(2-hydroxyethyl-d4)-L-cysteine dicyclohexylamine salt |
| CAS | 1331894-57-5 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C12 H23 N . C7 2H4 H9 N O4 S |
| Kvantitet | 5 mg |
| Synonym | (2R)-2-Acetamido-3-(1,1,2,2-tetradeuterio-2-hydroxyethyl)sulfanylpropanoic acid N-cyclohexylcyclohexanamine, N-Acetyl-3-[(2-hydroxyethyl-1,1,2,2-d4)thio]-L-alanine dicyclohexylammonium salt, N-Acetyl-S-(2-hydroxyethyl-1,1,2,2-d4)-L-cysteine dicyclohexylammonium salt, N-Acetyl-S-(2-hydroxyethyl)-L-cysteine-D4 (2-hydroxyethyl-1,1,2,2-D4) dicyclohexylammonium salt, N-Acetyl-S-(2-hydroxyethyl)cysteine-d4 dicyclohexylammonium salt, S-(2-Hydroxyethyl-d4)-N-acetylcysteine dicyclohexylammonium salt, S-(β-Hydroxyethyl)mercapturic-d4 acid dicyclohexylammonium salt, (2R)-2-Acetamido-3-(2-hydroxyethyl-1,1,2,2-d4)sulfanylpropanoic acid dicyclohexylamine |
| InChI formel | InChI=1S/C12H23N.C7H13NO4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5(10)8-6(7(11)12)4-13-3-2-9/h11-13H,1-10H2;6,9H,2-4H2,1H3,(H,8,10)(H,11,12)/t;6-/m.0/s1/i;2D2,3D2 |
| LEDER | [2H]C([2H])(O)C([2H])([2H])SC[C@H](NC(=O)C)C(=O)O.C1CCC(CC1)NC2CCCCC2 |
| IUPAC-namn | (2R)-2-acetamido-3-(1,1,2,2-tetradeuterio-2-hydroxyethyl)sulfanylpropanoic acid;N-cyclohexylcyclohexanamine |
| Molekylvikt (g/mol) | 392.59 |
| Visa mer |