Läs mer
N-Acetyl-S-(3,4-dihydroxybutyl)-L-cysteine (Mixture of Diastereomers), TRC
Produktkod. 30009939
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009939 10 mg
30006826 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009939 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA17371010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 144889-50-9
Molekylformel C9 H17 N O5 S
Molekylvikt (g/mol) 251.3
Synonym N-Acetyl-S-(3,4-Dihydroxybutyl)-L-Cysteine
IUPAC-namn (2R)-2-acetamido-3-(3,4-dihydroxybutylsulfanyl)propanoic acid
LEDER CC(=O)N[C@@H](CSCCC(O)CO)C(=O)O
Specifikationer
Kemiskt namn eller material N-Acetyl-S-(3,4-dihydroxybutyl)-L-cysteine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C9H17NO5S/c1-6(12)10-8(9(14)15)5-16-3-2-7(13)4-11/h7-8,11,13H,2-5H2,1H3,(H,10,12)(H,14,15)/t7?,8-/m0/s1
Kvantitet 10 mg
Formel vikt 251.0827
Procent renhet >95%