Läs mer
N-Acetyl-S-(3-hydroxypropyl-1-methyl)-L-cysteine Dicyclohexylammonium Salt(Mixture of Diastereomers), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Acetyl-S-(3-hydroxypropyl-1-methyl)-L-cysteine dicyclohexylammonium salt (mixture of diastereomers) |
| CAS | 33164-70-4 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C12 H23 N . C9 H17 N O4 S |
| Kvantitet | 10 mg |
| Synonym | L-Cysteine, N-acetyl-S-(3-hydroxy-1-methylpropyl)-, compd. with N-cyclohexylcyclohexanamine (1:1), Alanine, N-acetyl-3-[(3-hydroxy-1-methylpropyl)thio]-, compd. with dicyclohexylamine (1:1), L- (8CI), Dicyclohexylammonium 3-hydroxy-1-methylpropylmercapturic acid, N-Acetyl-S-(3-hydroxy-1-methylpropyl)-L-cysteine dicyclohexylammonium salt, Dicyclohexylammonium (2R)-2-acetamido-3-(4-hydroxybutan-2-ylthio)propanoic acid, N-Acetyl-S-(3-hydroxypropyl-1-methyl)-L-cysteine dicyclohexylammonium salt |
| InChI formel | InChI=1S/C12H23N.C9H17NO4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-6(3-4-11)15-5-8(9(13)14)10-7(2)12/h11-13H,1-10H2;6,8,11H,3-5H2,1-2H3,(H,10,12)(H,13,14)/t;6?,8-/m.0/s1 |
| LEDER | CC(CCO)SC[C@H](NC(=O)C)C(=O)O.C1CCC(CC1)NC2CCCCC2 |
| IUPAC-namn | (2R)-2-acetamido-3-(4-hydroxybutan-2-ylsulfanyl)propanoic acid;N-cyclohexylcyclohexanamine |
| Molekylvikt (g/mol) | 416.62 |
| Visa mer |