Läs mer
N-Acetyl-S-(3-hydroxypropyl)-L-cysteine Dicyclohexylamine Salt (unlabelled), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Acetyl-S-(3-hydroxypropyl)-L-cysteine dicyclohexylammonium salt |
| CAS | 14369-42-7 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C12 H23 N . C8 H15 N O4 S |
| Kvantitet | 0.5 mg |
| Synonym | L-Cysteine, N-acetyl-S-(3-hydroxypropyl)-, compd. with N-cyclohexylcyclohexanamine (1:1), Alanine, N-acetyl-3-[(3-hydroxypropyl)thio]-, compd. with dicyclohexylamine (1:1), L- (8CI), Alanine, N-acetyl-3-[(3-hydroxypropyl)thio]-, compd. with dicyclohexylamine (7CI), Dicyclohexylamine, compd. with N-acetyl-3-[(3-hydroxypropyl)thio]-L-alanine (1:1), N-Acetyl-S-(3-hydroxypropyl)cysteine dicyclohexylammonium salt, N-Acetyl-S-(3-hydroxypropyl)-L-cysteine dicyclohexylammonium salt |
| InChI formel | InChI=1S/C12H23N.C8H15NO4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-6(11)9-7(8(12)13)5-14-4-2-3-10/h11-13H,1-10H2;7,10H,2-5H2,1H3,(H,9,11)(H,12,13)/t;7-/m.0/s1 |
| LEDER | CC(=O)N[C@@H](CSCCCO)C(=O)O.C1CCC(CC1)NC2CCCCC2 |
| IUPAC-namn | (2R)-2-acetamido-3-(3-hydroxypropylsulfanyl)propanoic acid;N-cyclohexylcyclohexanamine |
| Molekylvikt (g/mol) | 402.59 |
| Visa mer |