Läs mer
N-Acetyl-S-benzyl-DL-cysteine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Acetyl-S-benzyl-DL-cysteine |
| CAS | 19538-71-7 |
| Molekylformel | C12H15NO3S |
| Kvantitet | 50 mg |
| Synonym | Cysteine, N-acetyl-S-(phenylmethyl)- (9CI, ACI), DL-Cysteine, N-acetyl-S-(phenylmethyl)- (ZCI), Alanine, N-acetyl-3-(benzylthio)-, DL- (8CI), N-Acetyl-S-(phenylmethyl)cysteine (ACI), 2-Acetamido-3-(benzylthio)propanoic acid, N-Acetyl-S-benzyl-DL-cysteine |
| InChI formel | InChI=1S/C12H15NO3S/c1-9(14)13-11(12(15)16)8-17-7-10-5-3-2-4-6-10/h2-6,11H,7-8H2,1H3,(H,13,14)(H,15,16) |
| LEDER | CC(=O)NC(CSCc1ccccc1)C(=O)O |
| IUPAC-namn | 2-acetamido-3-benzylsulfanylpropanoic acid |
| Molekylvikt (g/mol) | 253.31 |
| Formel vikt | 253.0773 |