Läs mer
N-Acetyl-S-methyl-L-cysteine, TRC
Produktkod. 30326102
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30326102 100 mg
1 options
Produktkod. 30326102 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA186640100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Acetyl-S-methyl-L-cysteine
CAS 16637-59-5
Molekylformel C6 H11 N O3 S
Kvantitet 100 mg
InChI formel InChI=1S/C6H11NO3S/c1-4(8)7-5(3-11-2)6(9)10/h5H,3H2,1-2H3,(H,7,8)(H,9,10)/t5-/m0/s1
LEDER CSC[C@H](NC(=O)C)C(=O)O
IUPAC-namn (2R)-2-acetamido-3-methylsulfanylpropanoic acid
Molekylvikt (g/mol) 177.22
Formel vikt 177.046