Läs mer
N-Acetyl Sulfamethoxazole, TRC
Produktkod. 17903635
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17903635 50 mg
17963535 25 mg
17912175 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17903635 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18788550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 21312-10-7
Molekylformel C12 H13 N3 O4 S
Molekylvikt (g/mol) 295.31
Synonym Acetanilide, 4'-[(5-methyl-3-isoxazolyl)sulfamoyl]- (6CI,7CI,8CI), N-[4-[[(5-Methyl-3-isoxazolyl)amino]sulfonyl]phenyl]acetamide, 4'-Acetyl-3-sulfa-5-methylisoxazole, Acetylsulfamethoxazole, N-Acetylsulfamethoxazole, N4-Acetylsulfamethoxazole, N4-Acetylsulfamethoxazole, N4-Acetylsulfisomezole, STX 606, Sulfisomezole-N4-acetate
IUPAC-namn N-[4-[(5-methyl-1,2-oxazol-3-yl)sulfamoyl]phenyl]acetamide
LEDER CC(=O)Nc1ccc(cc1)S(=O)(=O)Nc2cc(C)on2
Specifikationer
Kemiskt namn eller material Acetylsulfamethoxazole
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C12H13N3O4S/c1-8-7-12(14-19-8)15-20(17,18)11-5-3-10(4-6-11)13-9(2)16/h3-7H,1-2H3,(H,13,16)(H,14,15)
Formel vikt 295.0627
Procent renhet >95%