Läs mer
N-Acetyl Sulfapyridine, TRC
Produktkod. 30137159
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137159 50 mg
30134906 25 mg
30005767 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137159 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18791050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19077-98-6
Molekylformel C13 H13 N3 O3 S
Molekylvikt (g/mol) 291.33
Synonym Acetamide, N-[4-[(2-pyridinylamino)sulfonyl]phenyl]- (9CI, ACI), Acetanilide, 4'-(2-pyridylsulfamoyl)- (6CI, 7CI, 8CI), N-[4-[(2-Pyridinylamino)sulfonyl]phenyl]acetamide (ACI), Acetylsulfapyridine, N4-Acetylsulfapyridine, Sulfapyridine-N4-acetyl
IUPAC-namn N-[4-(pyridin-2-ylsulfamoyl)phenyl]acetamide
LEDER CC(=O)Nc1ccc(cc1)S(=O)(=O)Nc2ccccn2
Specifikationer
Kemiskt namn eller material N-Acetyl Sulfapyridine
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C13H13N3O3S/c1-10(17)15-11-5-7-12(8-6-11)20(18,19)16-13-4-2-3-9-14-13/h2-9H,1H3,(H,14,16)(H,15,17)
Formel vikt 291.0678
Procent renhet >95%