missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Acetyl-Val-Glu-Ile-Asp-p-nitroanilide

Colorimetric substrate for caspase-6

Brand:  Alfa Aesar™ J64992.MB

Additional Details : CAS Number : 189684-54-6 Weight : 0.00025kg

 View more versions of this product

Product Code. 15476679

  • 2410.37 SEK / 25mg


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Specifications

25mg
Lyophilized Solid
189684-54-6
636.65g/mol
Signal Transduction Reagent
−30° to −10°C
Colorimetric substrate for caspase-6
C28H40N6O11
Ac-VEID-pNA
Videos
SDS