missing translation for 'onlineSavingsMsg'
Läs mer

N-Acetyl Zonisamide, TRC

Produktkod. 30316076
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30316076 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30316076 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18980010MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-Acetyl Zonisamide
CAS 68936-43-6
Molekylformel C10 H10 N2 O4 S
Kvantitet 10 mg
Synonym Acetamide, N-[(1,2-benzisoxazol-3-ylmethyl)sulfonyl]-, 1,2-Benzisoxazole, acetamide deriv., N-[(1,2-Benzisoxazol-3-ylmethyl)sulfonyl]acetamide, N-Acetyl-3-(sulfamoylmethyl)-1,2-benzisoxazole, N-Acetyl Zonisamide
InChI formel InChI=1S/C10H10N2O4S/c1-7(13)12-17(14,15)6-9-8-4-2-3-5-10(8)16-11-9/h2-5H,6H2,1H3,(H,12,13)
LEDER CC(=O)NS(=O)(=O)Cc1noc2ccccc12
IUPAC-namn N-(1,2-benzoxazol-3-ylmethylsulfonyl)acetamide
Molekylvikt (g/mol) 254.26
Formel vikt 254.0361
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.