Läs mer
N-Acetylcysteine Amide, TRC
Produktkod. 17912475
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17912475 100 mg
17972775 25 mg
17983465 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17912475 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA172195100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38520-57-9
Molekylformel C5H10N2O2S
Molekylvikt (g/mol) 162.21
Synonym (2R)-2-(Acetylamino)-3-mercaptopropanamide; N-Acetyl-L-cysteinamide
IUPAC-namn (2R)-2-acetamido-3-sulfanylpropanamide
LEDER CC(=O)N[C@@H](CS)C(=O)N
Specifikationer
Kemiskt namn eller material N-Acetylcysteine Amide
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C5H10N2O2S/c1-3(8)7-4(2-10)5(6)9/h4,10H,2H2,1H3,(H2,6,9)(H,7,8)/t4-/m0/s1
Kvantitet 100 mg
Procent renhet >95%