Läs mer
Tocris Bioscience™ N-Acetylpuromycin
Downregulates SnoN and Ski protein expression; promotes TGF-β signaling
Brand: Tocris Bioscience™ 5679/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
N-Acetylpuromycin is a downregulates SnoN and Ski protein expression; promotes TGF-β signaling, independently of MAPK activation. Does not bind ribosomes or block protein synthesis.Specifikationer
| 3′-[[(2S)-2-(Acetylamino)-3-(4-methoxyphenyl)-1-oxopropyl]amino]-3′-deoxy-N,N-dimethyladenosine | |
| 22852-13-7 | |
| 10 mg |