Läs mer
N-Allyloxycarbonyl Moxifloxacin, TRC
Produktkod. 30318787
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30318787 50 mg
1 options
Produktkod. 30318787 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA56148050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Allyloxycarbonyl Moxifloxacin
CAS 1391053-44-3
Molekylformel C25H28FN3O6
Kvantitet 50 mg
Synonym 1-Cyclopropyl-8-methoxy-6-fluoro-1,4-dihydro-7-[(4aS,7aS)-N-allyloxycarbonyloctahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic Acid
InChI formel InChI=1S/C25H28FN3O6/c1-3-9-35-25(33)28-8-4-5-14-11-27(13-19(14)28)21-18(26)10-16-20(23(21)34-2)29(15-6-7-15)12-17(22(16)30)24(31)32/h3,10,12,14-15,19H,1,4-9,11,13H2,2H3,(H,31,32)/t14-,19+/m0/s1
LEDER COc1c(N2C[C@@H]3CCCN([C@@H]3C2)C(=O)OCC=C)c(F)cc4C(=O)C(=CN(C5CC5)c14)C(=O)O
IUPAC-namn 7-[(4aS,7aS)-1-prop-2-enoxycarbonyl-3,4,4a,5,7,7a-hexahydro-2H-pyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid
Molekylvikt (g/mol) 485.5
Formel vikt 485.1962