Läs mer
N-Aminomorpholine-4-carbothioamide, TRC
Produktkod. 30324040
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30324040 25 mg
30310384 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30324040 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61445825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Aminomorpholine-4-carbothioamide
CAS 6499-15-6
Molekylformel C5H11N3OS
Kvantitet 25 mg
InChI formel InChI=1S/C5H11N3OS/c6-7-5(10)8-1-3-9-4-2-8/h1-4,6H2,(H,7,10)
LEDER NNC(N1CCOCC1)=S
IUPAC-namn morpholine-4-carbothiohydrazide
Molekylvikt (g/mol) 161.22
Formel vikt 161.0623