missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-2'-deoxycytidine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7076531
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
4836-13-9
C16H17N3O5
331.33
MFCD00010115
MPSJHJFNKMUKCN-OUCADQQQSA-N
n4-benzoyl-2'-deoxycytidine, n-benzoyl-2'-deoxy-cytidine, n-benzoyl-2'-deoxycytidine, cytidine, n-benzoyl-2'-deoxy, cytidine,n-benzoyl-2'-deoxy, n-benzoyl-2-deoxy-cytidine, dc-bz, ksc590o7t, n-benzoyl-2/'-deoxycytidine
9797617
N-[1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide
OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O
This item is not returnable. View return policy
4836-13-9
C16H17N3O5
331.33
MFCD00010115
MPSJHJFNKMUKCN-OUCADQQQSA-N
n4-benzoyl-2'-deoxycytidine, n-benzoyl-2'-deoxy-cytidine, n-benzoyl-2'-deoxycytidine, cytidine, n-benzoyl-2'-deoxy, cytidine,n-benzoyl-2'-deoxy, n-benzoyl-2-deoxy-cytidine, dc-bz, ksc590o7t, n-benzoyl-2/'-deoxycytidine
9797617
N-[1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide
OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O

Reactant in the synthesis of 2', 5'-Dideoxycytidines and Other Derivatives of 2'-Deoxycytidine, N-Protected 2?-Deoxyribonucleosides(useful for oligonucleotide synthesis), 3?-, 5?-and 3?, 5?-di-O-crotonyl 2?-deoxynucleoside derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Packaging Glass bottle
Quantity 250 mg
Solubility Information Soluble in methanol (50mg/ml).
Formula Weight 331.33
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material N-Benzoyl-2′-deoxycytidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'