missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-L-tyrosine ethyl ester, 98%, Thermo Scientific Chemicals

Catalog Number 15450129
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3483-82-7
C18H19NO4
313.35
MFCD00002388
SRLROPAFMUDDRC-XISACWJONA-N
n-benzoyl-l-tyrosine ethyl ester, bz-tyr-oet, ethyl n-benzoyl-l-tyrosinate, btee, ethyl benzoyltyrosinate, benzoyltyrosine ethyl ester, benzoyl-l-tyrosine ethyl ester, s-ethyl 2-benzamido-3-4-hydroxyphenyl propanoate, benzoyl-tyr-oet, l-tyrosine, n-benzoyl-, ethyl ester
77033
ethyl (2S)-2-benzamido-3-(4-hydroxyphenyl)propanoate
CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
3483-82-7
C18H19NO4
313.35
MFCD00002388
SRLROPAFMUDDRC-XISACWJONA-N
n-benzoyl-l-tyrosine ethyl ester, bz-tyr-oet, ethyl n-benzoyl-l-tyrosinate, btee, ethyl benzoyltyrosinate, benzoyltyrosine ethyl ester, benzoyl-l-tyrosine ethyl ester, s-ethyl 2-benzamido-3-4-hydroxyphenyl propanoate, benzoyl-tyr-oet, l-tyrosine, n-benzoyl-, ethyl ester
77033
ethyl (2S)-2-benzamido-3-(4-hydroxyphenyl)propanoate
CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 118°C to 121°C
Beilstein 2223819
Formula Weight 313.35
Percent Purity 98%
Quantity 5 g
Physical Form Solid
Chemical Name or Material N-Benzoyl-L-tyrosine ethyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'