Läs mer
N-(Benzoyloxy)-N,a,a-trimethyl-benzeneethanamine, TRC
Produktkod. 30330234
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30330234 25 mg
1 options
Produktkod. 30330234 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB20852525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(Benzoyloxy)-N,alpha,alpha-trimethyl-benzeneethanamine
CAS 120583-17-7
Molekylformel C18H21NO2
Kvantitet 25 mg
Synonym O-Benzoyl-N-methyl-N-(2-methyl-1-phenylpropan-2-yl)hydroxylamine
InChI formel InChI=1S/C18H21NO2/c1-18(2,14-15-10-6-4-7-11-15)19(3)21-17(20)16-12-8-5-9-13-16/h4-13H,14H2,1-3H3
LEDER CN(OC(=O)c1ccccc1)C(C)(C)Cc2ccccc2
IUPAC-namn [methyl-(2-methyl-1-phenylpropan-2-yl)amino] benzoate
Molekylvikt (g/mol) 283.36
Formel vikt 283.1572