Läs mer
N-Benzy N,N-Didesmethyl Trimebutine, TRC
Produktkod. 30343652
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30343652 25 mg
1 options
Produktkod. 30343652 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB27569025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Benzy N,N-Didesmethyl Trimebutine
CAS 1246817-32-2
Molekylformel C27H31NO5
Kvantitet 25 mg
Synonym 3,4,5-Trimethoxybenzoic Acid 2-Phenyl-2-[(phenylmethyl)amino]butyl Ester
InChI formel InChI=1S/C27H31NO5/c1-5-27(22-14-10-7-11-15-22,28-18-20-12-8-6-9-13-20)19-33-26(29)21-16-23(30-2)25(32-4)24(17-21)31-3/h6-17,28H,5,18-19H2,1-4H3
LEDER CCC(COC(=O)c1cc(OC)c(OC)c(OC)c1)(NCc2ccccc2)c3ccccc3
IUPAC-namn [2-(benzylamino)-2-phenylbutyl] 3,4,5-trimethoxybenzoate
Molekylvikt (g/mol) 449.54
Formel vikt 449.2202