missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-3-(tert-butyldimethylsilanyloxymethyl)piperid-4-one, 95%, Thermo Scientific™

Catalog Number p-3522593
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
325486-37-1
C19H31NO2Si
333.55
MFCD09065031
WOYQEVXRQBRUAL-UHFFFAOYNA-N
1-benzyl-3-tert-butyldimethylsilyl oxy methyl piperidin-4-one, n-benzy 3-tert-butyldimethylsilanyloxymethyl piperid-4-one, 1-benzyl-3-tert-butyldimethylsilyloxymethyl-piperidin-4-one, n-benzyl-3-tert-butyldimethylsilanyloxymethyl piperid-4-one, 1-benzyl-3-tert-butyl dimethyl silyl oxy methyl piperidin-4-one, 1-benzyl-3-tert-butyl dimethylsilyloxy methyl piperidin-4-one, 1-benzyl-3-tert-butyldimethylsilyloxy methyl piperidin-4-one, 4-piperidinone,3-1,1-dimethylethyl dimethylsilyl oxy methyl-1-phenylmethyl, 1-benzyl-3-tert-butyldimethylsilyl oxy methyl-4-oxopiperidine, 1-benzyl-4-oxopiperidin-3-yl methoxy tert-butyldimethylsilane
22495722
1-benzyl-3-[[tert-butyl(dimethyl)silyl]oxymethyl]piperidin-4-one
CC(C)(C)[Si](C)(C)OCC1CN(CC2=CC=CC=C2)CCC1=O
This item is not returnable. View return policy
325486-37-1
C19H31NO2Si
333.55
MFCD09065031
WOYQEVXRQBRUAL-UHFFFAOYNA-N
1-benzyl-3-tert-butyldimethylsilyl oxy methyl piperidin-4-one, n-benzy 3-tert-butyldimethylsilanyloxymethyl piperid-4-one, 1-benzyl-3-tert-butyldimethylsilyloxymethyl-piperidin-4-one, n-benzyl-3-tert-butyldimethylsilanyloxymethyl piperid-4-one, 1-benzyl-3-tert-butyl dimethyl silyl oxy methyl piperidin-4-one, 1-benzyl-3-tert-butyl dimethylsilyloxy methyl piperidin-4-one, 1-benzyl-3-tert-butyldimethylsilyloxy methyl piperidin-4-one, 4-piperidinone,3-1,1-dimethylethyl dimethylsilyl oxy methyl-1-phenylmethyl, 1-benzyl-3-tert-butyldimethylsilyl oxy methyl-4-oxopiperidine, 1-benzyl-4-oxopiperidin-3-yl methoxy tert-butyldimethylsilane
22495722
1-benzyl-3-[[tert-butyl(dimethyl)silyl]oxymethyl]piperidin-4-one
CC(C)(C)[Si](C)(C)OCC1CN(CC2=CC=CC=C2)CCC1=O
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 333.54
Percent Purity 95%
Chemical Name or Material N-Benzyl-3-(tert-butyldimethylsilanyloxymethyl)piperid-4-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'