Läs mer
N-Benzyl Ciprofloxacin Benzyl Ester, TRC
Produktkod. 30348388
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30348388 50 mg
1 options
Produktkod. 30348388 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB23370050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Benzyl Ciprofloxacin Benzyl Ester
CAS 1391052-64-4
Molekylformel C31H30FN3O3
Kvantitet 50 mg
Synonym 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(4-benzyl-1-piperazinyl)-3-quinolinecarboxylic Acid Benzyl Ester
InChI formel InChI=1S/C31H30FN3O3/c32-27-17-25-28(18-29(27)34-15-13-33(14-16-34)19-22-7-3-1-4-8-22)35(24-11-12-24)20-26(30(25)36)31(37)38-21-23-9-5-2-6-10-23/h1-10,17-18,20,24H,11-16,19,21H2
LEDER Fc1cc2C(=O)C(=CN(C3CC3)c2cc1N4CCN(Cc5ccccc5)CC4)C(=O)OCc6ccccc6
IUPAC-namn benzyl 7-(4-benzylpiperazin-1-yl)-1-cyclopropyl-6-fluoro-4-oxoquinoline-3-carboxylate
Molekylvikt (g/mol) 511.59
Formel vikt 511.2271